missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Neurturin (Histidine-tagged) Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4902
Molecular Weight 14 kDa (monomer)
Name Neurturin (Histidine-tagged)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
387NE025 R&D Systems
RDS387NE025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $298.51
387NE025CF R&D Systems
RDS387NE025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $298.51
Specifications

Description

Source: E. coli-derived, Ala96-Val197
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 4902
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
14 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
4902
Neurturin (Histidine-tagged)
25μg
Human