Please login to your online account to display your discounted pricing

R&D Systems Human Neuregulin-1 Isoform SMDF Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3084
Molecular Weight 33 kDa
Name Neuregulin-1 Isoform SMDF
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

378SM025

 
r&d systems
RDS378SM025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

378SM025CF

 
r&d systems
RDS378SM025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3084
Molecular Weight 33 kDa
Name Neuregulin-1 Isoform SMDF
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Met1-Glu296, Leu117-Glu296 & Ala162-Glu296, each with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 33 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3084
Regulatory Status: RUO