missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Neuregulin-1 alpha/NRG1 alpha EGF Domain Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3084
Molecular Weight 7 kDa
Name Neuregulin-1 alpha/NRG1 alpha EGF Domain
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
296HR050 R&D Systems
RDS296HR050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
296HR050CF R&D Systems
RDS296HR050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: E. coli-derived, Ser177-Lys241
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3084
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3084
Neuregulin-1 alpha/NRG1 alpha EGF Domain
50μg
Human