Please login to your online account to display your discounted pricing

R&D Systems Human Netrin-4 Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 59277
Molecular Weight 69 kDa
Name Netrin-4
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1254N4025CF

 
r&d systems
RDS1254N4025CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Each for $419.81

1254N4025

 
r&d systems
RDS1254N4025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 59277
Molecular Weight 69 kDa
Name Netrin-4
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Val26-Lys628 , with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 69 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Entrez Gene ID: 59277
Regulatory Status: RUO