missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human MSP/MST1 (Cys672Ala) Recombinant Protein

$366.12 - $366.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4485
Molecular Weight 53 kDa ( alpha-chain), 25 kDa (beta-chain)
Name MSP/MST1 (Cys672Ala)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4306MS010CF R&D Systems
RDS4306MS010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $366.12
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Gln19-Arg483 (alpha) & Val484-Gly711 (Cys672Ala) (beta)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 53 kDa ( alpha-chain), 25 kDa (beta-chain)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4485
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
53 kDa ( alpha-chain), 25 kDa (beta-chain)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
4485
MSP/MST1 (Cys672Ala)
10μg
Human