missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human MSP/MST1 β Chain Recombinant Protein

$442.13 - $442.13

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4485
Molecular Weight 25 kDa
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Name Price Quantity  
Catalog Number Mfr. No. Formulation Name Price Quantity  
635MS025 R&D Systems
RDS635MS025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. MSP/MST1 β Chain Each for $442.13
635MS025CF R&D Systems
RDS635MS025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. MSP/MST1 y Chain Each for $442.13
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Val484-Gly711
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 4485
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
25 kDa
25μg
Human
4485
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes