missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human/Mouse Wnt-5a Recombinant Protein

$456.25 - $456.25

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7474
Molecular Weight 38 kDa
Name Wnt-5a
Purity 80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
645WN010 R&D Systems
RDS645WN010
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. Each for $456.25
645WN010CF R&D Systems
RDS645WN010CF
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. Each for $456.25
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Gln38-Lys380
Purity: 80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 38 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Entrez Gene ID: 7474
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
38 kDa
80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7474
Wnt-5a
10μg
Human/Mouse