Please login to your online account to display your discounted pricing

R&D Systems Human/Mouse/Rat/Canine/Equine BDNF Recombinant Protein

$298.51 - $866.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 627
Molecular Weight 13.5 kDa
Name BDNF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

248BD025

 
r&d systems
RDS248BD025
Lyophilized from a 0.2μm filtered solution in Sodium Citrate and NaCl with BSA as a carrier protein. 25μg Each for $866.87

248BD025CF

 
r&d systems
RDS248BD025CF
Lyophilized from a 0.2μm filtered solution in Sodium Citrate and NaCl. 25μg Each for $866.87

248BD005

 
r&d systems
RDS248BD005
Lyophilized from a 0.2μm filtered solution in Sodium Citrate and NaCl with BSA as a carrier protein. 5μg Each for $298.51

248BD005CF

 
r&d systems
RDS248BD005CF
Lyophilized from a 0.2μm filtered solution in Sodium Citrate and NaCl. 5μg Each for $298.51
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 627
Molecular Weight 13.5 kDa
Name BDNF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, His129-Arg247
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Citrate and NaCl.
Entrez Gene ID: 627
Regulatory Status: RUO