missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human/Mouse Indian Hedgehog/Ihh (C28II), N-Terminus Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3549
Molecular Weight 20 kDa
Name Indian Hedgehog/Ihh (C28II), N-Terminus
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1705HH025 R&D Systems
RDS1705HH025
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose and with BSA as a carrier protein. Each for $416.79
1705HH025CF R&D Systems
RDS1705HH025CF
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose. Each for $416.79
Specifications

Description

Source: E. coli-derived, Cys28-Gly202 (Cys28Ile-Ile), with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose.
Entrez Gene ID: 3549
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3549
Indian Hedgehog/Ihh (C28II), N-Terminus
25μg
Human/Mouse