Please login to your online account to display your discounted pricing

R&D Systems Human MIS/AMH Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 268
Molecular Weight 11.7 kDa (monomer)
Name MIS/AMH
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1737MS010

 
r&d systems
RDS1737MS010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $419.81

1737MS010CF

 
r&d systems
RDS1737MS010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 268
Molecular Weight 11.7 kDa (monomer)
Name MIS/AMH
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Human

Source: E. coli-derived, Ala453-Arg560
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 11.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 268
Regulatory Status: RUO