Please login to your online account to display your discounted pricing

R&D Systems Human Midkine Recombinant Protein

$389.76 - $520.12

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 4192
Molecular Weight 13.3 kDa
Name Midkine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

258MD050

 
r&d systems
RDS258MD050
50μg Each for $520.12

258MD010

 
r&d systems
RDS258MD010
10μg Each for $389.76
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 4192
Molecular Weight 13.3 kDa
Name Midkine
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Lys23-Asp143
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4192
Regulatory Status: RUO