Please login to your online account to display your discounted pricing

R&D Systems Human Methylated Ubiquitin Recombinant Protein

$70.18 - $287.11

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a solution in deionized water.
Gene ID (Entrez) 7314
Molecular Weight 8.7 kDa
Name Methylated Ubiquitin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

U50101M

 
r&d systems
RDSU50101M
1mg Each for $70.18

U50105M

 
r&d systems
RDSU50105M
5mg Each for $287.11
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a solution in deionized water.
Gene ID (Entrez) 7314
Molecular Weight 8.7 kDa
Name Methylated Ubiquitin
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 8.7 kDa
Formulation: Lyophilized from a solution in deionized water.
Entrez Gene ID: 7314
Regulatory Status: RUO