Please login to your online account to display your discounted pricing

R&D Systems Human Methylated Ubiquitin Recombinant Protein


Manufacturer: r&d systems  RDSU50105M

 View more versions of this product

Catalog No. U50105M

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 8.7 kDa
Formulation Lyophilized from a solution in deionized water.
Quantity 5mg
Name Methylated Ubiquitin
Gene ID (Entrez) 7314
Source Human
Conjugate Unlabeled
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes