missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human MD-1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 9450
Name MD-1
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
925MD050 R&D Systems
RDS925MD050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 18.2 kDa Each for $428.87
925MD050CF R&D Systems
RDS925MD050CF
Lyophilized from a 0.2μm filtered solution in PBS. 32.5 kDa Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gly21-Ser162
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 32.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 9450
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
MD-1
50μg
Human
9450
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes