Please login to your online account to display your discounted pricing

R&D Systems Human MBL Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4153
Molecular Weight 24 kDa (monomer)
Name MBL
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

2307MB050

 
r&d systems
RDS2307MB050
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and BSA as a carrier protein. Each for $428.87

2307MB050CF

 
r&d systems
RDS2307MB050CF
Supplied as a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4153
Molecular Weight 24 kDa (monomer)
Name MBL
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Glu21-Ile248
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24 kDa (monomer)
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4153
Regulatory Status: RUO