missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Lymphotoxin α1/β2 Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4049
Molecular Weight 18 kDa (LT alpha), 18.5 kDa (LTbeta)
Name Lymphotoxin α1/β2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
678LY010 R&D Systems
RDS678LY010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $441.90
678LY010CF R&D Systems
RDS678LY010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $441.90
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Leu35-Leu205 (LT alpha) & Leu54-Gly244 (LT beta), both with an N-terminal Met & Thr41-Leu205 (LT alpha)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa (LT alpha), 18.5 kDa (LTbeta)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4049
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa (LT alpha), 18.5 kDa (LTbeta)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
4049
Lymphotoxin α1/β2
10μg
Human