missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human LIGHT/TNFSF14 Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8740
Molecular Weight 20.9 kDa
Name LIGHT/TNFSF14
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
664LI025 R&D Systems
RDS664LI025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $415.83
664LI025CF R&D Systems
RDS664LI025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $415.83
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Asp74-Val240, with a N-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8740
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
8740
LIGHT/TNFSF14
25μg
Human