missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human LIF Recombinant Protein

$366.12 - $1,265.57

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3976
Molecular Weight 19.6 kDa
Name LIF
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
7734LF025 R&D Systems
RDS7734LF025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $366.12
7734LF100CF R&D Systems
RDS7734LF100CF
Lyophilized from a 0.2μm filtered solution in PBS. 100μg Each for $1,265.57
7734LF100 R&D Systems
RDS7734LF100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 100μg Each for $1,265.57
7734LF025CF R&D Systems
RDS7734LF025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $366.12
Specifications

Description

Source: E. coli-derived, Pro24-Phe202
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3976
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
19.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3976
LIF
Yes