Please login to your online account to display your discounted pricing

R&D Systems Human Leptin/OB Recombinant Protein

$272.44 - $676.55

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Gene ID (Entrez) 3952
Molecular Weight 16 kDa
Name Leptin/OB
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

398LP01M

 
r&d systems
RDS398LP01M
1mg Each for $272.44

398LP05M

 
r&d systems
RDS398LP05M
5mg Each for $676.55
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Gene ID (Entrez) 3952
Molecular Weight 16 kDa
Name Leptin/OB
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Val22-Cys167, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Entrez Gene ID: 3952
Regulatory Status: RUO