missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human LDL R Recombinant Protein

$429.46 - $429.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3949
Molecular Weight 85.6 kDa & 66.8 kDa
Name LDL R
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2148LD025 R&D Systems
RDS2148LD025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $429.46
2148LD025CF R&D Systems
RDS2148LD025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $429.46
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala22-Arg788, with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 85.6 kDa & 66.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3949
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
85.6 kDa & 66.8 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3949
LDL R
25μg
Human