Please login to your online account to display your discounted pricing

R&D Systems Human LBP (Histidine-tagged) Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3929
Molecular Weight 51.7 kDa
Name LBP (Histidine-tagged)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

6445LP025

 
r&d systems
RDS6445LP025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81

6445LP025CF

 
r&d systems
RDS6445LP025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3929
Molecular Weight 51.7 kDa
Name LBP (Histidine-tagged)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Ala26-Val481 with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 51.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3929
Regulatory Status: RUO