missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Latent TGF-beta 1 Recombinant Protein

$376.73 - $1,041.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7040
Molecular Weight 41.3 kDa (monomer)
Name Latent TGF-beta 1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
299LT005 R&D Systems
RDS299LT005
Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein. 5μg Each for $376.73
299LT025CF R&D Systems
RDS299LT025CF
Supplied as a 0.2μm filtered solution in PBS and Glycerol. 25μg Each for $1,041.55
299LT005CF R&D Systems
RDS299LT005CF
Supplied as a 0.2μm filtered solution in PBS and Glycerol. 5μg Each for $376.73
299LT025 R&D Systems
RDS299LT025
Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein. 25μg Each for $1,041.55
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Leu30-Ser390
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 41.3 kDa (monomer)
Formulation: Supplied as a 0.2μm filtered solution in PBS and Glycerol.
Entrez Gene ID: 7040
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
41.3 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
7040
Latent TGF-beta 1
Yes