Please login to your online account to display your discounted pricing

R&D Systems Human L-Selectin/CD62L Recombinant Protein


Manufacturer: r&d systems  RDSADP2050

 View more versions of this product

Catalog No. ADP2050

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 33.1 kDa
Name L-Selectin/CD62L
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
Gene ID (Entrez) 6402
Quantity 50μg
Source Human