missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Kininostatin Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3827
Molecular Weight 10.5 kDa
Name Kininostatin
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1396KN050 R&D Systems
RDS1396KN050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
1396KN050CF R&D Systems
RDS1396KN050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: E. coli-derived, Lys438-Ser531
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3827
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
10.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3827
Kininostatin
50μg
Human