Please login to your online account to display your discounted pricing

R&D Systems Human Insulin R/CD220 Recombinant Protein

$432.57 - $432.57

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3643
Molecular Weight 82.9 kDa ( alpha subunit), 22.9 kDa (beta subunit)
Name Insulin R/CD220
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1544IR050

 
r&d systems
RDS1544IR050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $432.57

1544IR050CF

 
r&d systems
RDS1544IR050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $432.57
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3643
Molecular Weight 82.9 kDa ( alpha subunit), 22.9 kDa (beta subunit)
Name Insulin R/CD220
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, His28-Arg750 (alpha subunit) & Ser751-Lys944 with a C-terminal 10-His tag (beta subunit)
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 82.9 kDa ( alpha subunit), 22.9 kDa (beta subunit)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3643
Regulatory Status: RUO