Please login to your online account to display your discounted pricing

R&D Systems Human IL-9 Recombinant Protein

$407.05 - $1,167.56

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3578
Molecular Weight 14.1 kDa
Name IL-9
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

209IL050

 
r&d systems
RDS209IL050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $1,167.56

209IL050CF

 
r&d systems
RDS209IL050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $1,167.56

209IL010CF

 
r&d systems
RDS209IL010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $407.05
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3578
Molecular Weight 14.1 kDa
Name IL-9
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gln19-Ile144
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3578
Regulatory Status: RUO