missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-9 R Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3581
Molecular Weight 27.4 kDa
Name IL-9 R
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
290RNS025 R&D Systems
RDS290RNS025
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. Each for $428.87
290RNS025CF R&D Systems
RDS290RNS025CF
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Val40-Pro270, with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 27.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Entrez Gene ID: 3581
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
27.4 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3581
IL-9 R
25μg
Human