missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-7 Recombinant Protein

$298.51 - $932.05

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3574
Molecular Weight 17 kDa
Name IL-7
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
207IL025 R&D Systems
RDS207IL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $932.05
207IL005 R&D Systems
RDS207IL005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $298.51
207IL005CF R&D Systems
RDS207IL005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $298.51
207IL025CF R&D Systems
RDS207IL025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $932.05
Specifications

Description

Source: E. coli-derived, Asp26-His177, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3574
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3574
IL-7
Yes