Please login to your online account to display your discounted pricing

R&D Systems Human IL-6 Recombinant Protein, (HEK293-expressed)

$483.61 - $483.61

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3569
Molecular Weight 20.8 kDa
Name IL-6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

7270IL025

 
r&d systems
RDS7270IL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $483.61

7270IL025CF

 
r&d systems
RDS7270IL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $483.61
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3569
Molecular Weight 20.8 kDa
Name IL-6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Human embryonic kidney cell, HEK293-derived, Met1-Met212
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3569
Regulatory Status: RUO