Please login to your online account to display your discounted pricing

R&D Systems Human IL-6 Recombinant Protein

$292.21 - $4,338.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3569
Molecular Weight 20.3 kDa
Name IL-6
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

206IL001MGC

 
r&d systems
RDS206IL001MGCF
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. 1mg Each for $4,338.46

206IL010CF

 
r&d systems
RDS206IL010CF
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. 10μg Each for $292.21

206IL010

 
r&d systems
RDS206IL010
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. 10μg Each for $292.21

206IL050CF

 
r&d systems
RDS206IL050CF
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. 50μg Each for $930.21

206IL050

 
r&d systems
RDS206IL050
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. 50μg Each for $930.21
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3569
Molecular Weight 20.3 kDa
Name IL-6
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Pro29-Met212
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Entrez Gene ID: 3569
Regulatory Status: RUO