missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-6 R alpha Recombinant Protein

$480.13 - $480.13

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3570
Molecular Weight 38 kDa
Name IL-6 R alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
227SR025 R&D Systems
RDS227SR025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $480.13
227SR025CF R&D Systems
RDS227SR025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $480.13
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Leu20-Asp358
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 38 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3570
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
38 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3570
IL-6 R alpha
25μg
Human