missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-5 Recombinant Protein

$226.76 - $695.50

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3567
Name IL-5
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity  
205IL005CF R&D Systems
RDS205IL005CF
Supplied as a 0.2μm filtered solution in PBS. 13.1 kDa (monomer) 5μg Each for $226.76
205IL025 R&D Systems
RDS205IL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 13 kDa (monomer) 25μg Each for $695.50
205IL025CF R&D Systems
RDS205IL025CF
Supplied as a 0.2μm filtered solution in PBS. 13.1 kDa (monomer) 25μg Each for $695.50
205IL005 R&D Systems
RDS205IL005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 13 kDa (monomer) 5μg Each for $226.76
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (stably transfected)-derived, Ile20-Ser134
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.1 kDa (monomer)
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3567
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-5
Yes
3567
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human