missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-4 Recombinant Protein, CHO-expressed)

$272.44 - $272.44

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3565
Molecular Weight 15.0 kDa
Name IL-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6507IL010CF R&D Systems
RDS6507IL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $272.44
6507IL010 R&D Systems
RDS6507IL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $272.44
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, His25-Ser153
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.0 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3565
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.0 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3565
IL-4
10μg
Human