missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-4 R alpha Recombinant Protein

$556.14 - $556.14

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3566
Molecular Weight 24 kDa
Name IL-4 R alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2304R025 R&D Systems
RDS2304R025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $556.14
2304R025CF R&D Systems
RDS2304R025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $556.14
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gly24-His232
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3566
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
24 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3566
IL-4 R alpha
25μg
Human