Please login to your online account to display your discounted pricing

R&D Systems Human IL-37b/IL-1F7b (aa 46-218) Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 27178
Molecular Weight 19.4 kDa
Name IL-37b/IL-1F7b (aa 46-218)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

7585IL025CF

 
r&d systems
RDS7585IL025CF
Lyophilized from a 0.2μm filtered solution in PBS and DTT. Each for $419.81

7585IL025

 
r&d systems
RDS7585IL025
Lyophilized from a 0.2μm filtered solution in PBS and DTT with BSA as a carrier protein. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 27178
Molecular Weight 19.4 kDa
Name IL-37b/IL-1F7b (aa 46-218)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Val46-Asp218
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and DTT.
Entrez Gene ID: 27178
Regulatory Status: RUO