missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-36Ra/IL-1F5 Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 26525
Molecular Weight 17 kDa
Name IL-36Ra/IL-1F5
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1275IL025 R&D Systems
RDS1275IL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $441.90
1275IL025CF R&D Systems
RDS1275IL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $441.90
Specifications

Description

Source: E. coli-derived, Val2-Asp155
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 26525
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
26525
IL-36Ra/IL-1F5
25μg
Human