missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-36 beta/IL-1F8 (aa 5-157) Recombinant Protein

For use in cell or tissue culture applications.

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 27177
Molecular Weight 17.2 kDa
Name IL-36 beta/IL-1F8 (aa 5-157)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6834IL010 R&D Systems
RDS6834IL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

  • Source: E. coli-derived, Arg5-Glu157
  • Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: 17.2 kDa
  • Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
  • Entrez Gene ID: 27177
  • Regulatory Status: RUO
Description & Specifications

Specifications

Unlabeled
17.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
27177
IL-36 beta/IL-1F8 (aa 5-157)
10μg
Human