missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-36 alpha/IL-1F6 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 27179
Molecular Weight 17.7 kDa
Name IL-36 alpha/IL-1F6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1078IL025 R&D Systems
RDS1078IL025
Lyophilized from a 0.2μm filtered solution in PBS, NaCl and Tween™ 20 with BSA as a carrier protein. Each for $428.87
1078IL025CF R&D Systems
RDS1078IL025CF
Lyophilized from a 0.2μm filtered solution in PBS, NaCl and Tween™ 20. Each for $428.87
Specifications

Description

Source: E. coli-derived, Met1-Phe158
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, NaCl and Tween™ 20.
Entrez Gene ID: 27179
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.7 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
27179
IL-36 alpha/IL-1F6
25μg
Human