missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-3 R alpha/CD123 Recombinant Protein

$572.26 - $572.26

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3563
Molecular Weight 33 kDa
Name IL-3 R alpha/CD123
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
301R3025 R&D Systems
RDS301R3025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $572.26
301R3025CF R&D Systems
RDS301R3025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $572.26
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Lys20-Arg305
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 33 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3563
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
33 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3563
IL-3 R alpha/CD123
25μg
Human