missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-28B/IFN-lambda 3 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 282617
Molecular Weight 20.1 kDa
Name IL-28B/IFN-lambda 3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5259IL025CF R&D Systems
RDS5259IL025CF
Lyophilized from a 0.2μm filtered solution in PBS, NaCl and EDTA. Each for $416.79
5259IL025 R&D Systems
RDS5259IL025
Lyophilized from a 0.2μm filtered solution in PBS, NaCl and EDTA with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Arg30-Val200, with the variant of Lys74Arg and a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, NaCl and EDTA.
Entrez Gene ID: 282617
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
282617
IL-28B/IFN-lambda 3
25μg
Human