missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-26/AK155 Dimer Recombinant Protein

$429.46 - $429.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 55801
Molecular Weight 17.7 kDa (monomer)
Name IL-26/AK155 Dimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1870IL010 R&D Systems
RDS1870IL010
Supplied as a 0.2μm filtered solution in NaH2PO4, NaCl and Glycerol with BSA as a carrier protein. Each for $429.46
1870IL010CF R&D Systems
RDS1870IL010CF
Supplied as a 0.2μm filtered solution in NaH2PO4, NaCl and Glycerol. Each for $429.46
Specifications

Description

Source: E. coli-derived, Lys22-Gln171, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.7 kDa (monomer)
Formulation: Supplied as a 0.2μm filtered solution in NaH2PO4, NaCl and Glycerol.
Entrez Gene ID: 55801
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.7 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
55801
IL-26/AK155 Dimer
10μg
Human