Please login to your online account to display your discounted pricing

R&D Systems Human IL-23 Recombinant Protein

$407.05 - $407.05

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 51561
Name IL-23
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity    

1290IL010

 
r&d systems
RDS1290IL010
Lyophilized from a 0.2μm filtered solution in MES, NaCl and EDTA with BSA as a carrier protein. 55.0 kDa (IL-23 single chain); 36.5 kDa (p40) & 18.5 kDa (p19) Each for $407.05

1290IL010CF

 
r&d systems
RDS1290IL010CF
Supplied as a 0.2μm filtered solution in MES, NaCl and EDTA. 55.0 kDa (IL-23 single chain), 36.5 kDa (p40), 18.5 kDa (p19) Each for $407.05
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 51561
Name IL-23
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ile23-Ser328 (p40) & Ala21-Pro189 (p19)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 55.0 kDa (IL-23 single chain), 36.5 kDa (p40), 18.5 kDa (p19)
Formulation: Supplied as a 0.2μm filtered solution in MES, NaCl and EDTA.
Entrez Gene ID: 51561
Regulatory Status: RUO