Please login to your online account to display your discounted pricing

R&D Systems Human IL-20 Recombinant Protein

$427.46 - $427.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 50604
Molecular Weight 17.6 kDa
Name IL-20
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1102IL025

 
r&d systems
RDS1102IL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $427.46

1102IL025CF

 
r&d systems
RDS1102IL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $427.46
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 50604
Molecular Weight 17.6 kDa
Name IL-20
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Leu25-Glu176, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 50604
Regulatory Status: RUO