Please login to your online account to display your discounted pricing

R&D Systems Human IL-2 Recombinant Protein

$253.93 - $797.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3558
Molecular Weight 15 kDa
Name IL-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

202IL010CF

 
r&d systems
RDS202IL010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $253.93

202IL010

 
r&d systems
RDS202IL010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $253.93

202IL050CF

 
r&d systems
RDS202IL050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $797.51

202IL050

 
r&d systems
RDS202IL050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $797.51
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3558
Molecular Weight 15 kDa
Name IL-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Ala21-Thr153, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 3558
Regulatory Status: RUO