Please login to your online account to display your discounted pricing

R&D Systems Human IL-2 R beta Recombinant Protein

$246.37 - $741.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3560
Molecular Weight 24.8 kDa
Name IL-2 R beta
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

2242B005CF

 
r&d systems
RDS2242B005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $246.37

2242B025CF

 
r&d systems
RDS2242B025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $741.73

2242B025

 
r&d systems
RDS2242B025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $741.73

2242B005

 
r&d systems
RDS2242B005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $246.37
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3560
Molecular Weight 24.8 kDa
Name IL-2 R beta
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala27-Asp239
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3560
Regulatory Status: RUO