Please login to your online account to display your discounted pricing

R&D Systems Human IL-2 (Cys145Ser) Recombinant Protein


Manufacturer: r&d systems  RDS1081IL020

 View more versions of this product

Catalog No. 1081IL020

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 15.5 kDa
Recombinant Yes
Formulation Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein.
Name IL-2 (Cys145Ser)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 20μg
Source Human
Conjugate Unlabeled
Gene ID (Entrez) 3558