missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-2 (Cys145Ser) Recombinant Protein

$285.48 - $285.48

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3558
Molecular Weight 15.5 kDa
Name IL-2 (Cys145Ser)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1081IL020 R&D Systems
RDS1081IL020
Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. Each for $285.48
1081IL020CF R&D Systems
RDS1081IL020CF
Supplied as a 0.2μm filtered solution in Sodium Acetate. Each for $285.48
Specifications

Description

Source: E. coli-derived, Ala21-Thr153 (Cys145Ser), with and without an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.5 kDa
Formulation: Supplied as a 0.2μm filtered solution in Sodium Acetate.
Entrez Gene ID: 3558
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3558
IL-2 (Cys145Ser)
20μg
Human