missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-17F Recombinant Protein, (NS0-expressed)

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 112744
Molecular Weight 15.7 kDa (monomer)
Name IL-17F
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1335INS025 R&D Systems
RDS1335INS025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $376.73
1335INS025C R&D Systems
RDS1335INS025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $376.73
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Arg31-Gln163, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 112744
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.7 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
112744
IL-17F
25μg
Human