Please login to your online account to display your discounted pricing

R&D Systems Human IL-17E/IL-25 Recombinant Protein


Manufacturer: r&d systems  RDS1258IL025

 View more versions of this product

Catalog No. 1258IL025

  • / Each
 

Description & Specifications

Specifications

Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Name IL-17E/IL-25
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 64806
Molecular Weight 17 kDa (monomer)
Structural Form Disulfide-linked homodimer
Conjugate Unlabeled
Source Human
Recombinant Yes
Quantity 25μg