missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human IL-17D Recombinant Protein

$290.11 - $290.11

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 53342
Molecular Weight 20 kDa (monomer)
Name IL-17D
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1504IL025CF R&D Systems
RDS1504IL025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $290.11
1504IL025 R&D Systems
RDS1504IL025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $290.11
Specifications

Description

Source: E. coli-derived, Ala18-Pro202
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 53342
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
53342
IL-17D
25μg
Human